Learn More
Electron Microscopy Sciences Digitonin, Crystal, Reagent 1 GR
SDP
Supplier:  Electron Microscopy Sciences 13260
Encompass_Preferred
CAS #11024-24-1 C₅₆H₉₂O₂₉C₅₆H₉₂O₂₉ F.W. 1229.34
An additive which stabilizes cholesterol 0.2% digitonin in buffer added to Glutaraldehyde fixative may help to stabilize and retain the fine structural localization of free cholesterol and cholesterol esters. See Okros, I Histochemie 13-91, 1968. Windaus, A. Hoop Seyler's Z. Physio. Chem. 65:110, 1910. J. Cell Bio., 40, 802, 1969. or Temkin, Robert J. (1993); Microscopy Research and Tech. 26:260-271.
Catalog No. 50-980-361