Learn More
Abcam Digitonin, detergent for solubilizing receptors and cellular membranes, 1G
Catalog No. 502583356
Encompass_Preferred
MW 1229.3 Da. Digitonin, detergent for solubilizing receptors and cellular membranes. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3356 Supplier Abcam Supplier No. AB1415011G
MW 1229.3 Da. Digitonin, detergent for solubilizing receptors and cellular membranes. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement