Learn More
Selleck Chemical LLC Digitonin-E1321-100MG
SDP
Catalog No. 503137266
Encompass_Preferred
Digitonin a steroidal saponin and glycoside obtained from Digitalis purpurea has strong lytic activity on various cell membranes with an IC50 of 15 1 M on sheep RBC membranes
Catalog No. 50-313-7266 Supplier Selleck Chemical LLC Supplier No. E1321100MG
Digitonin a steroidal saponin and glycoside obtained from Digitalis purpurea has strong lytic activity on various cell membranes with an IC50 of 15 1 M on sheep RBC membranes