Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digitonin, MP Biomedicals™

Catalog No. MP215948091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215948091 1 g
MP215948050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215948091 Supplier MP Biomedicals, Inc Supplier No. 0215948091
Add to Cart
Add to Cart

Permeabilizes plasma membranes of eukaryotic cells. Digitonin, MP Biomedicals is a mild nonionic detergent that complexes with membrane cholesterol and other unconjugated β-hydroxysterols. It is used to solubilize receptors and permeabilize cellular

Digitonin has following uses:

  • Cholesterol determination
  • Use of digitonin-permeabilized adipocytes for cAMP studies
  • Solubilization of membrane proteins
  • The water soluble form can be used for the isolation of mitotic chromosomes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11024-24-1
Molecular Formula C56H92O29
Molecular Weight (g/mol) 1229.32
MDL Number MFCD00077729
InChI Key UVYVLBIGDKGWPX-QYLWGQLPSA-N
Synonym digitonin
PubChem CID 102004607
IUPAC Name (2S,3R,4S,5S,6R)-2-{[(2S,3R,4S,5S,6R)-2-{[(2S,3R,4S,5R,6R)-2-{[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(1'R,2R,2'S,3'S,4'R,7'S,8'R,9'S,12'S,13'S,15'R,16'R,18'S)-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosane]-3',15'-dioloxy]oxan-3-yl]oxy}-5-hydroxy-6-(hydroxymethyl)-4-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C[C@H]1[C@H]2[C@@H](O[C@]11CCC(C)CO1)[C@@H](O)[C@H]1[C@@H]3CC[C@H]4C[C@@H](O[C@@H]5O[C@H](CO)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O[C@@H]7OC[C@@H](O)[C@H](O)[C@H]7O)[C@H]6O[C@@H]6O[C@H](CO)[C@H](O)[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)C[C@]4(C)[C@H]3CC[C@]21C

Specifications

Physical Form White Powder
Melting Point 230°C to 240°C (decomposition)(literature)
Quantity 1 g
Specific Rotation [α]20/D −54° (c=2.8, methanol) (Lit.)
Solubility Information Soluble in boiling water (1g/10mL). Heat for 15 minutes at 98°C. Cool and place at 4°C for 3 hrs and filter off any precipitate. Soluble in absolute alcohol (1g/57mL) and 95% alcohol (1g).
Formula Weight 1229.3
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) <0.5mM (20–25°C) (Lit.)
Recommended Storage Store at Room Temperature (15°C to 30°C), dessicate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.