Learn More
Abcam Digitonin Solution, 1ML
Catalog No. 502703812
Encompass_Preferred
Digitonin Solution (ab300022) is a 5% solution of Digitonin in deionised water.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3812 Supplier Abcam Supplier No. AB3000221ML
Digitonin Solution (ab300022) is a 5% solution of Digitonin in deionised water.
The product is subject to the following: Abcam Restricted Use Statement