Learn More
Alkali Scientific Digitonin, Water Soluble, 1 G
SDP
Catalog No. 5000661847
Encompass_Preferred
Digitonin is a water-soluble detergent used for membrane permeabilization and the extraction of cellular components in molecular and cellular biology research. The 1g quantity is suitable for laboratories conducting large-scale experiments or for use in ongoing studies involving membrane protein research. Digitonin interacts with cholesterol to form pores in cell membranes, allowing researchers to isolate intracellular components such as proteins, enzymes, and organelles. It is widely used in experiments focused on enzyme assays, protein purification, and the study of membrane dynamics. This reagent is indispensable in cell biology, proteomics, and biochemistry labs for the extraction of membrane-bound proteins and other intracellular entities.
Catalog No. 50-006-61847 Supplier Alkali Scientific Supplier No. DIG002CP
Digitonin is a water-soluble detergent used for membrane permeabilization and the extraction of cellular components in molecular and cellular biology research. The 1g quantity is suitable for laboratories conducting large-scale experiments or for use in ongoing studies involving membrane protein research. Digitonin interacts with cholesterol to form pores in cell membranes, allowing researchers to isolate intracellular components such as proteins, enzymes, and organelles. It is widely used in experiments focused on enzyme assays, protein purification, and the study of membrane dynamics. This reagent is indispensable in cell biology, proteomics, and biochemistry labs for the extraction of membrane-bound proteins and other intracellular entities.