Learn More
Crescent Chemical Co Inc DIGITONIN WATER SOLUBLE
SDP
Catalog No. 50247513
Encompass_Preferred
Digitonin water soluble research grade Turbidity: 1 g is suspended in 20 ml water and dissolved by heating to 95 - 98 °C for 15 minutes. Size - 250MG Storage Conditions - +15 °C TO +30 °C Catalog Number - 19551.01 CAS 11024-24-1
Catalog No. 50-247-513 Supplier Crescent Chemical Co Inc Supplier No. SE19551.01
Digitonin water soluble research grade Turbidity: 1 g is suspended in 20 ml water and dissolved by heating to 95 - 98 °C for 15 minutes. Size - 250MG Storage Conditions - +15 °C TO +30 °C Catalog Number - 19551.01 CAS 11024-24-1