Learn More
Alkali Scientific Digitonin, Water Soluble, 500 Milligrams
SDP
Catalog No. 5000661125
Encompass_Preferred
Digitonin is a water-soluble compound used primarily in membrane permeabilization for research applications. Available in a 500mg format, Digitonin is widely used in cell biology to permeabilize cell membranes, allowing researchers to study intracellular components or deliver compounds directly into cells. It is particularly useful in protein extraction, cellular signaling studies, and in assays that require the breakdown of membrane integrity without damaging the cells entirely. This high-purity product facilitates effective permeabilization while maintaining cellular viability, making it essential for research on cellular processes, membrane proteins, and intracellular interactions.
Catalog No. 50-006-61125 Supplier Alkali Scientific Supplier No. DIG003CP
Digitonin is a water-soluble compound used primarily in membrane permeabilization for research applications. Available in a 500mg format, Digitonin is widely used in cell biology to permeabilize cell membranes, allowing researchers to study intracellular components or deliver compounds directly into cells. It is particularly useful in protein extraction, cellular signaling studies, and in assays that require the breakdown of membrane integrity without damaging the cells entirely. This high-purity product facilitates effective permeabilization while maintaining cellular viability, making it essential for research on cellular processes, membrane proteins, and intracellular interactions.