Learn More
Cayman Chemical Digitoxigenin, Purity: ≥95%, CAS: 143-62-4, 10 mg
SDP
Catalog No. 502968153
Encompass_Preferred
A cardenolide and aglycone constituent of digitoxin that elicits cardiac contraction and cardiotonic effects by inhibiting the Na+/K+ ATPase; inhibits protein synthesis and has been examined for use as an antitumor compound
Catalog No. 50-296-8153 Supplier Cayman Chemical Supplier No. 1822910 MG
A cardenolide and aglycone constituent of digitoxin that elicits cardiac contraction and cardiotonic effects by inhibiting the Na+/K+ ATPase; inhibits protein synthesis and has been examined for use as an antitumor compound