Learn More
Glas Col LLC Control
SDP
Catalog No. NC9815145
Encompass_Preferred
Dgitrol featurse a digital dual output display for set point and process temperature indication. The automatic-tuning feature ensures increased temperature accuracy with minimum overshoot. Easy set-up and simple input commands make our controls very "user friendly". Display: LED Process/Setpoint. Digital auto-temperature display of setpoint and process temperatures. Universal input. 120V, 1800W. 0-750 degrees C range.
Catalog No. NC9815145 Supplier Glas Col LLC Supplier No. 104A PL612
Dgitrol featurse a digital dual output display for set point and process temperature indication. The automatic-tuning feature ensures increased temperature accuracy with minimum overshoot. Easy set-up and simple input commands make our controls very "user friendly". Display: LED Process/Setpoint. Digital auto-temperature display of setpoint and process temperatures. Universal input. 120V, 1800W. 0-750 degrees C range.