Learn More
SCP Science DigiTUBE, 100ml, RL/No Caps | 300/pack
Catalog No. NC1404143
Encompass
RackLock DigiTUBEs graduated at every 10ml mark along with readings at 25, 50, 75 and 100ml levels. They are Class 'A' calibrated at the 50ml and 100ml mark. Does not include caps.
Catalog No. NC1404143 Supplier SCP Science Supplier No. 010501262
RackLock DigiTUBEs graduated at every 10ml mark along with readings at 25, 50, 75 and 100ml levels. They are Class 'A' calibrated at the 50ml and 100ml mark. Does not include caps.