Learn More
SCP Science SCP Science DigiTUBE Autosampler Tubes, 15 mL Volume, Ultra Low Metal Content, Non-Sterile
Catalog No. NC0734136
Encompass
The DigiTUBE Autosampler Tubes are designed for accurate sample handling in scientific applications. Made from virgin polypropylene, these 15 mL tubes excel in preventing contamination with ultra-low metal content, ensuring reliable results. Their robust construction withstands common acids, alkalis, and solvents, enhancing laboratory efficiency by fitting standard autosampler racks. Ideal for various laboratory procedures, these non-sterile tubes are packaged in quantities of 1000, offering great volume for extensive usage.
Catalog No. NC0734136 Supplier SCP Science Supplier No. 010515607
The DigiTUBE Autosampler Tubes are designed for accurate sample handling in scientific applications. Made from virgin polypropylene, these 15 mL tubes excel in preventing contamination with ultra-low metal content, ensuring reliable results. Their robust construction withstands common acids, alkalis, and solvents, enhancing laboratory efficiency by fitting standard autosampler racks. Ideal for various laboratory procedures, these non-sterile tubes are packaged in quantities of 1000, offering great volume for extensive usage.