Learn More
Perkin Elmer US LLC DigiTUBEs 100 mL non-RackLock, Includes Caps, Qty. 300
Catalog No. 502103760
Encompass_Preferred
All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.
Catalog No. 50-210-3760 Supplier Perkin Elmer US LLC Supplier No. N9308066
All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.