Learn More
Perkin Elmer US LLC DigiTUBEs 15 mL with RackLock with Caps, Pkg. 540
Catalog No. 502105413
Encompass_Preferred
All-in-one tubes eliminate the need for five separate vessels: Digestion beakers Volumetric flasks Graduated cylinders Autosampler tubes Storage containers.
Catalog No. 50-210-5413 Supplier Perkin Elmer US LLC Supplier No. N9308713
All-in-one tubes eliminate the need for five separate vessels: Digestion beakers Volumetric flasks Graduated cylinders Autosampler tubes Storage containers.