Learn More
SCP Science DigiTUBEs 50 mL Non-RackLock, High Acid Resistance, Ultra-Low Leachable Metals
Catalog No. NC0891766
Encompass
The SCP Science DigiTUBEs 50 mL Non-RackLock are advanced disposable digestion vessels ideal for analytical laboratories. Their robust virgin polypropylene construction ensures high visibility and acid resistance. Designed with ultra-low leachable metal content and certified accuracy, these multi-purpose tubes enhance laboratory efficiency by reducing the need for multiple vessels, streamlining workflows, and lowering costs. Suitable for various applications, they offer a significant advantage over traditional alternatives.
Catalog No. NC0891766 Supplier SCP Science Supplier No. 010500264
The SCP Science DigiTUBEs 50 mL Non-RackLock are advanced disposable digestion vessels ideal for analytical laboratories. Their robust virgin polypropylene construction ensures high visibility and acid resistance. Designed with ultra-low leachable metal content and certified accuracy, these multi-purpose tubes enhance laboratory efficiency by reducing the need for multiple vessels, streamlining workflows, and lowering costs. Suitable for various applications, they offer a significant advantage over traditional alternatives.