Learn More
Enzo Life Sciences Digoxigenin-UTP, alkali-stable (250nmol)
SDP
Catalog No. 502013894
Encompass_Preferred
Digoxigenin-UTP can replace UTP in reactions catalyzed by T3, T7 or SP6 RNA polymerases. The digoxigenin-labeled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridization, sequencing, and genome analysis. The transcription reaction produces multiple RNA copies of the DNA template(s) during a short incubation period. RNA probes offer higher target specificity and greater sensitivity than the corresponding DNA-DNA hybrids. The single-stranded RNA probes offer selectivity unavailable with double-stranded DNA probes, because they are strand-specific. Furthermore, RNA probes hybridize much more efficiently to target molecules than DNA probes because there is no self-hybridization. Alternative name: Digoxigenin-11-UTP. Concentration: 10mM. Quantity: 25µl. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.
Catalog No. 50-201-3894 Supplier Enzo Life Sciences Supplier No. ENZNUC1140250
Digoxigenin-UTP can replace UTP in reactions catalyzed by T3, T7 or SP6 RNA polymerases. The digoxigenin-labeled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridization, sequencing, and genome analysis. The transcription reaction produces multiple RNA copies of the DNA template(s) during a short incubation period. RNA probes offer higher target specificity and greater sensitivity than the corresponding DNA-DNA hybrids. The single-stranded RNA probes offer selectivity unavailable with double-stranded DNA probes, because they are strand-specific. Furthermore, RNA probes hybridize much more efficiently to target molecules than DNA probes because there is no self-hybridization. Alternative name: Digoxigenin-11-UTP. Concentration: 10mM. Quantity: 25µl. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.