Learn More
Apexbio Technology LLC Digoxigenin-11-dUTP 25ul (1 mM)
SDP
Catalog No. 5000088802
Encompass_Preferred
Digoxigenin-11-dUTP is a modified nucleotide analog designed for incorporation into DNA or cDNA by enzymatic processes substituting for natural dTTP The digoxigenin moiety attached via an 11-atom linker to the C5-position of the uridine base facilitates recognition by anti-digoxigenin antibodies labeled with horseradish peroxidase alkaline phosphatase or fluorophores Digoxigenin-11-dUTP is enzymatically incorporated using methods such as PCR amplification nick translation primer extension 3 -end labeling and reverse transcription Based on these properties Digoxigenin-11-dUTP is primarily utilized in probe-based assays sequencing and fluorescent or chromogenic in situ hybridization (ISH) applications
Catalog No. 50-000-88802 Supplier Apexbio Technology LLC Supplier No. B79520.025
Digoxigenin-11-dUTP is a modified nucleotide analog designed for incorporation into DNA or cDNA by enzymatic processes substituting for natural dTTP The digoxigenin moiety attached via an 11-atom linker to the C5-position of the uridine base facilitates recognition by anti-digoxigenin antibodies labeled with horseradish peroxidase alkaline phosphatase or fluorophores Digoxigenin-11-dUTP is enzymatically incorporated using methods such as PCR amplification nick translation primer extension 3 -end labeling and reverse transcription Based on these properties Digoxigenin-11-dUTP is primarily utilized in probe-based assays sequencing and fluorescent or chromogenic in situ hybridization (ISH) applications