Learn More
Sigma Aldrich Fine Chemicals Biosciences Digoxigenin 11 dUTP alkali
Catalog No. 502914718
Encompass
Digoxigenin-11-dUTP alkali-stable is provided in 1 mM tetralithium salt SOLUTIONon.
Catalog No. 50-291-4718 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11558706910
Digoxigenin-11-dUTP alkali-stable is provided in 1 mM tetralithium salt SOLUTIONon.