Learn More
Sigma Aldrich Fine Chemicals Biosciences Digoxigenin 11 UTP
Catalog No. 502905020
Encompass
Digoxigenin (DIG) -11-uridine triphosphate (UTP) is provided as a SOLUTIONon of the tetralithium salt in 3.5mM (200nmol) or 10mM (250nmol) concentration. It is used as a substrate for SP6 T3 and T7 RNA polymerases. It can replace UTP in the in vitro transcription reaction for DIG labeling of RNA in a ratio of 3565%.
Catalog No. 50-290-5020 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11209256910
Digoxigenin (DIG) -11-uridine triphosphate (UTP) is provided as a SOLUTIONon of the tetralithium salt in 3.5mM (200nmol) or 10mM (250nmol) concentration. It is used as a substrate for SP6 T3 and T7 RNA polymerases. It can replace UTP in the in vitro transcription reaction for DIG labeling of RNA in a ratio of 3565%.