Learn More
Apexbio Technology LLC Digoxigenin-11-UTP 25ul (10 mM)
SDP
Catalog No. 503107353
Encompass_Preferred
Digoxigenin-11-UTP is a modified nucleoside triphosphate used for labeling RNA probes It incorporates a digoxigenin label attached to uridine triphosphate (UTP) allowing it to be incorporated into RNA molecules during in vitro transcription reactions Digoxigenin is a non-radioactive label widely utilized in molecular biology experiments especially in situ hybridization (ISH) and Northern blotting RNA probes labeled with Digoxigenin-11-UTP exhibit high sensitivity and low background noise providing clear detection results Additionally the digoxigenin label binds specifically to anti-digoxigenin antibodies making the detection process both straightforward and highly efficient Digoxigenin-11-UTP serves as an essential tool for studying gene expression and RNA localization
Catalog No. 50-310-7353 Supplier Apexbio Technology LLC Supplier No. B79530.025
Digoxigenin-11-UTP is a modified nucleoside triphosphate used for labeling RNA probes It incorporates a digoxigenin label attached to uridine triphosphate (UTP) allowing it to be incorporated into RNA molecules during in vitro transcription reactions Digoxigenin is a non-radioactive label widely utilized in molecular biology experiments especially in situ hybridization (ISH) and Northern blotting RNA probes labeled with Digoxigenin-11-UTP exhibit high sensitivity and low background noise providing clear detection results Additionally the digoxigenin label binds specifically to anti-digoxigenin antibodies making the detection process both straightforward and highly efficient Digoxigenin-11-UTP serves as an essential tool for studying gene expression and RNA localization