Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxigenin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018060 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8060 500 mg
11-101-8880 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8060 Supplier MilliporeSigma Supelco Supplier No. D9026500MG
Add to Cart
Add to Cart

Digoxigenin is a therapeutic drug belonging to the group of cardiac glycosides, widely used in the management of congestive heart failure and other cardiac diseases.

Featured Industries: Environmental and Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 1672-46-4
Physical Form Neat
Linear Formula C23H34O5
MDL Number MFCD00871861
Melting Point 222°C
Synonym 3 beta,12 beta,14 beta,21-Tetrahydroxy-20(22)-norcholenic acid lactone; 3 beta,12 beta,14-Trihydroxy-5 beta,20(22)-cardenolide; 5 beta,20(22)-Cardenolide-3 beta,12 beta,14-triol; Lanadigigenin
Type Analytical Standard
Quantity 500 mg
Formula Weight 390.51
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.