Learn More
Cayman Chemical DIgoxIgenIn BIsdIgItoxosID 1mg
SDP
Supplier:  Cayman Chemical 353561 MG
Encompass_Preferred
A Na+/K+-ATPase inhibitor; an active metabolite of digoxin; inhibits the Na+/K+-ATPase isoforms α1β1 and α2β1 (Kis = 196 and 74 nM, respectively)
Catalog No. 50-291-3395