Learn More
Cayman Chemical DIgoxIgenIn BIsdIgItoxosID
SDP
Catalog No. 502913757
Encompass_Preferred
A Na+/K+-ATPase inhibitor; an active metabolite of digoxin; inhibits the Na+/K+-ATPase isoforms α1β1 and α2β1 (Kis = 196 and 74 nM, respectively)
Catalog No. 50-291-3757 Supplier Cayman Chemical Supplier No. 35356500 UG
A Na+/K+-ATPase inhibitor; an active metabolite of digoxin; inhibits the Na+/K+-ATPase isoforms α1β1 and α2β1 (Kis = 196 and 74 nM, respectively)