Learn More
Medchemexpress LLC Digoxigenin NHS ester | 129273-26-3 | MFCD02093394 | 99.0% | 658.78 g/mol | C35H50N2O10 | 10 MG
SDP
Catalog No. 5000285881
Encompass_Preferred
Digoxigenin NHS ester is an activated N-hydroxysuccinimide (NHS) ester that conjugates the digoxigenin hapten to primary amines on proteins, peptides, or oligonucleotides under mild conditions. Supplied as a purified powder for biomolecule labeling and detection workflows, it is commonly used to prepare non-radioactive probes for in situ hybridization and immunodetection.
  • High purity, typically 99.0%.
  • Reacts with primary amines under mild conditions.
  • Suitable for labeling proteins and oligonucleotides.
  • Supplied as a stable powder with defined storage recommendations.
  • Enables non-radioactive detection in molecular assays.
Catalog No. 50-002-85881 Supplier Medchemexpress LLC Supplier No. HY15550010MG
Digoxigenin NHS ester is an activated N-hydroxysuccinimide (NHS) ester that conjugates the digoxigenin hapten to primary amines on proteins, peptides, or oligonucleotides under mild conditions. Supplied as a purified powder for biomolecule labeling and detection workflows, it is commonly used to prepare non-radioactive probes for in situ hybridization and immunodetection.
  • High purity, typically 99.0%.
  • Reacts with primary amines under mild conditions.
  • Suitable for labeling proteins and oligonucleotides.
  • Supplied as a stable powder with defined storage recommendations.
  • Enables non-radioactive detection in molecular assays.