Learn More
Aat Bioquest Digoxigenin NHS ester (1 mg)
SDP
Catalog No. 502249392
Encompass_Preferred
Digoxigenin DIG is a hapten a small molecule with high antigenicity which is used in many biological detections similarly to other popular haptens such as 24-Dinitrophenol DNP and biotin. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-224-9392 Supplier Aat Bioquest Supplier No. 3500
Digoxigenin DIG is a hapten a small molecule with high antigenicity which is used in many biological detections similarly to other popular haptens such as 24-Dinitrophenol DNP and biotin. This catalog is for 1 mg.