Learn More
Enzo Life Sciences Digoxigenin polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2660100
Encompass_Preferred
Immunogen: Digoxin. Source: Purified from sheep serum. Species reactivity: Species independent. Specificity: Recognizes Digoxin and Digoxigenin. Applications: ELISA, WB TUNEL, ISH (in situ hybridization). Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-2811