Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digoxin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111020499 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0499 100 mg
11-101-8693 1 g
11-101-8014 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-0499 Supplier MilliporeSigma Supelco Supplier No. D6003100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Digoxin
CAS 20830-75-5
Shipping Condition Ambient
Physical Form Neat
Linear Formula C41H64O14
Technique Gas Chromatography
MDL Number MFCD00003674
Synonym 12 beta-Hydroxydigitoxin
Type Analytical Standard
Solvent or Matrix Neat
Industry Type Drug Analysis
Molecular Weight (g/mol) 780.94
Quantity 100 mg
Formula Weight 780.94
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.