Learn More
Selleck Chemical LLC Digoxin100mg
SDP
Supplier:  Selleck Chemical LLC S4290100MG
Encompass_Preferred
Digoxin (NSC 95100) is a classical Na,K-ATPase inhibitor, with selectivity for the ?2?3 isoform over the common ?1?1 isoform, used in the treatment of atrial fibrillation and heart failure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3141