Learn More
Cayman Chemical Digoxind3 GC MS Standards
SDP
Supplier:  Cayman Chemical 10010657500 UG
Encompass_Preferred
An internal standard for the quantification of digoxin by GC- or LC-MS
Catalog No. 50-288-8545