Learn More
Cayman Chemical Digoxin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 127299-95-0, 1 mg
SDP
Catalog No. 502889601
Encompass_Preferred
An internal standard for the quantification of digoxin by GC- or LC-MS
Catalog No. 50-288-9601 Supplier Cayman Chemical Supplier No. 100106571 MG
An internal standard for the quantification of digoxin by GC- or LC-MS