Learn More
AVANTOR MASTERFLEX (VWR LLC) DIGTL DR .02-100RPM 115/230VAC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0755130
Encompass
Masterflex L/S Digital Drive, Computer-Compatible, 0.02 to 100 rpm; 115/230 VAC
Catalog No. NC2183894