Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGTL HEAT ULTSC BTH 230/230V
Catalog No. NC3721480
Encompass_Preferred
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX3800H-E, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 230/240V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank
Catalog No. NC3721480 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952338R
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX3800H-E, Capacity: 5.7L (1.5 gal.), Dimensions Interior: 29 x 15 x 15cm (11.5 x 6 x 6 in.), Voltage: 230/240V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover and drain, Plastic Housing, Stainless Steel Tank