Learn More
Cole-Parmer Cole-Parmer SHP-200D-L-C-120 Digital Stirring Hot Plate, Ceramic, 12" x 12"; 120 VAC
Catalog No. NC3789796 Shop All Cole-Parmer Products
Encompass_Preferred
-Powerful magnets for strong magnetic coupling. -Microprocessor controlled, dual thermocouples ensure accurate temperature control. -Large plate area accommodates one vessel up to 15 L. -Easy-to-use temperature and stirring control dials. -Hot warning light remains active even when switched off, but still connected to the mains supply. -BioCote® antimicrobial protection is built in for a cleaner, more hygienic product.
Catalog No. NC3789796 Supplier Cole-Parmer Supplier No. 0480777
-Powerful magnets for strong magnetic coupling. -Microprocessor controlled, dual thermocouples ensure accurate temperature control. -Large plate area accommodates one vessel up to 15 L. -Easy-to-use temperature and stirring control dials. -Hot warning light remains active even when switched off, but still connected to the mains supply. -BioCote® antimicrobial protection is built in for a cleaner, more hygienic product.