Learn More
Enzo Life Sciences DIGX Mouse anti-digoxigenin linker (Ready-to-Use) (6ml)
SDP
Catalog No. 502012842
Encompass_Preferred
The DIGX Mouse anti-digoxigenin linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity  of Enzo's DIGX  HPV probes. Although the DIGX Mouse anti-digoxigenin linker was created to enhance Enzo's DIGX probes, it will recognize any digoxigenin associated nucleotide or macromolecule. Clone: HY-A.1. Isotype: IgG1. Source: Purified IgG fraction of mouse serum. Species reactivity: Species independent . Applications: ISH (in situ hybridization). Formulation: Liquid. Ready-to-use in PBS containing stabilizers and preservatives.
Catalog No. 50-201-2842 Supplier Enzo Life Sciences Supplier No. ENZABS3046000
The DIGX Mouse anti-digoxigenin linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity  of Enzo's DIGX  HPV probes. Although the DIGX Mouse anti-digoxigenin linker was created to enhance Enzo's DIGX probes, it will recognize any digoxigenin associated nucleotide or macromolecule. Clone: HY-A.1. Isotype: IgG1. Source: Purified IgG fraction of mouse serum. Species reactivity: Species independent . Applications: ISH (in situ hybridization). Formulation: Liquid. Ready-to-use in PBS containing stabilizers and preservatives.