Learn More
Enzo Life Sciences DIGX Rabbit anti-digoxigenin linker (Ready-to-Use) (6ml)
SDP
Catalog No. 502012841
Encompass_Preferred
The DIGX Rabbit  anti-digoxigenin linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity of Enzo's DIGX HPV probes. Although the DIGX Rabbit anti-digoxigenin linker was created to enhance Enzo's DIGX probes, it will recognize any digoxigenin associated nucleotide or macromolecule. Isotype: IgG. Source: Purified IgG fraction of rabbit serum. Species reactivity: Species independent . Applications: ISH (in situ hybridization). Formulation: Liquid. Ready-to-use in PBS containing stabilizers and preservatives.
Catalog No. 50-201-2841 Supplier Enzo Life Sciences Supplier No. ENZABS3036000
The DIGX Rabbit  anti-digoxigenin linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity of Enzo's DIGX HPV probes. Although the DIGX Rabbit anti-digoxigenin linker was created to enhance Enzo's DIGX probes, it will recognize any digoxigenin associated nucleotide or macromolecule. Isotype: IgG. Source: Purified IgG fraction of rabbit serum. Species reactivity: Species independent . Applications: ISH (in situ hybridization). Formulation: Liquid. Ready-to-use in PBS containing stabilizers and preservatives.