Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diheptylamine 98.0+%, TCI America™
SDP

Catalog No. D26755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26755G Supplier TCI America Supplier No. D26755G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2470-68-0
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.41
MDL Number MFCD00027309
InChI Key NJWMENBYMFZACG-UHFFFAOYSA-N
PubChem CID 75580
IUPAC Name diheptylamine
SMILES CCCCCCCNCCCCCCC

Specifications

Color Colorless
Boiling Point 148°C
Quantity 5 g
UN Number 2735
Formula Weight 213.41
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diheptylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.