Learn More
Medchemexpress LLC Diheptylviologen dibromide | 6159-05-3 | MFCD00011997 | 99.2% | 514.38 g/mol | C24H38Br2N2 | 1 ML
SDP
Catalog No. 5000211109
Encompass_Preferred
DHBP dibromide (Diheptylviologen dibromide) is a research-use compound that inhibits calcium release and exhibits muscle relaxant activity. It is provided as a 10 mM solution in DMSO (1 mL vial) for in vitro experiments and should not be used for human or veterinary applications. The compound is identified by CAS 6159-05-3, molecular formula C24H38Br2N2, and a molecular weight of approximately 514.38 g/mol.
  • Inhibits calcium release from intracellular stores.
  • Exhibits muscle relaxant activity useful for physiological studies.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity (99.17%) suitable for research applications.
  • Intended for in vitro research use only.
Catalog No. 50-002-11109 Supplier Medchemexpress LLC Supplier No. HY10123710MM 1ML
DHBP dibromide (Diheptylviologen dibromide) is a research-use compound that inhibits calcium release and exhibits muscle relaxant activity. It is provided as a 10 mM solution in DMSO (1 mL vial) for in vitro experiments and should not be used for human or veterinary applications. The compound is identified by CAS 6159-05-3, molecular formula C24H38Br2N2, and a molecular weight of approximately 514.38 g/mol.
  • Inhibits calcium release from intracellular stores.
  • Exhibits muscle relaxant activity useful for physiological studies.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity (99.17%) suitable for research applications.
  • Intended for in vitro research use only.