Learn More
Medchemexpress LLC Diheptylviologen dibromide | 6159-05-3 | MFCD00011997 | 99.2% | 514.38 g/mol | C24H38Br2N2 | 500 MG
SDP
Catalog No. 5000211526
Encompass_Preferred
DHBP dibromide (Diheptylviologen dibromide) is a research-use chemical that functions as an inhibitor of calcium release and a muscle relaxant, provided as a high-purity reagent for laboratory studies in ion-channel biology and muscle physiology.
  • Inhibits calcium release in cellular and biochemical assays.
  • Acts as a muscle relaxant for physiological research applications.
  • High purity suitable for analytical and experimental workflows.
  • Available as a solid in common laboratory quantities, including 500 mg.
  • Documented chemical identity with CAS number and formula for traceability.
Catalog No. 50-002-11526 Supplier Medchemexpress LLC Supplier No. HY101237500MG
DHBP dibromide (Diheptylviologen dibromide) is a research-use chemical that functions as an inhibitor of calcium release and a muscle relaxant, provided as a high-purity reagent for laboratory studies in ion-channel biology and muscle physiology.
  • Inhibits calcium release in cellular and biochemical assays.
  • Acts as a muscle relaxant for physiological research applications.
  • High purity suitable for analytical and experimental workflows.
  • Available as a solid in common laboratory quantities, including 500 mg.
  • Documented chemical identity with CAS number and formula for traceability.