Learn More
TARGETMOL CHEMICALS INC DIHEXA 5MG
SDP
Catalog No. 502576320
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dihexa (Hexanoyl-Tyr-Ile-Ahx-NH2) is an activator of the hepatocyte growth factor/c-Met (HGF/c-Met) systemit binds to HGF (Kd = 65 pM)and an analog of the peptide angiotensin IV. purity: 98%
Catalog No. 50-257-6320 Supplier TARGETMOL CHEMICALS INC Supplier No. T73765MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dihexa (Hexanoyl-Tyr-Ile-Ahx-NH2) is an activator of the hepatocyte growth factor/c-Met (HGF/c-Met) systemit binds to HGF (Kd = 65 pM)and an analog of the peptide angiotensin IV. purity: 98%