Learn More
Medchemexpress LLC Dihexadecyl hydrogen 1g
SDP
Catalog No. 5000206857
Encompass_Preferred
Dihexadecyl hydrogen phosphateIt is an organic compound belonging to phospholipids It s often used as an emulsifier which means it helps mix two substances together that don t usually mix well such as oil and water Dihexadecyl hydrogen phosphateIt has several applications in the food industry especially in the production of processed foods where it improves texture and stability Additionally it has applications in the pharmaceutical industry where it can be used ain the generation of micelles liposomes and other types of artificial membranes
Catalog No. 50-002-06857 Supplier Medchemexpress LLC Supplier No. HYW0995471G
Dihexadecyl hydrogen phosphateIt is an organic compound belonging to phospholipids It s often used as an emulsifier which means it helps mix two substances together that don t usually mix well such as oil and water Dihexadecyl hydrogen phosphateIt has several applications in the food industry especially in the production of processed foods where it improves texture and stability Additionally it has applications in the pharmaceutical industry where it can be used ain the generation of micelles liposomes and other types of artificial membranes