Learn More
Cayman Chemical DIhexadecyl Phosphate 1g
SDP
Catalog No. 502933025
Encompass_Preferred
A synthetic phospholipid; has been used to impart a negative charge to neutral liposomes; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including niosomes for drug delivery
Catalog No. 50-293-3025 Supplier Cayman Chemical Supplier No. 257271 G
A synthetic phospholipid; has been used to impart a negative charge to neutral liposomes; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including niosomes for drug delivery