Learn More
Cayman Chemical Dihexadecyl Phosphate, Purity: ≥90%, CAS: 2197-63-9, 5 g
SDP
Catalog No. 502936158
Encompass_Preferred
A synthetic phospholipid; has been used to impart a negative charge to neutral liposomes; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including niosomes for drug delivery
Catalog No. 50-293-6158 Supplier Cayman Chemical Supplier No. 257275 G
A synthetic phospholipid; has been used to impart a negative charge to neutral liposomes; has been used in the generation of micelles, liposomes, and other types of artificial membranes, including niosomes for drug delivery