Learn More
Medchemexpress LLC Dihexyl phthalate-3,4,5,6-d4 | 1015854-55-3 | 98.2% | 338.47 | 10 MG
SDP
Catalog No. 5000452908
Encompass_Preferred
Dihexyl phthalate-3,4,5,6-d4 is the deuterium labeled Dihexyl phthalate-3,4,5,6. Dihexyl phthalate is one of the commonly used phthalate esters in various plastics and consumer products. It is classified as a priority pollutant and an endocrine disruptor, capable of inducing reactive oxygen species (ROS) accumulation, promoting inflammation, and leading to increased apoptosis and inflammation-related gene expression levels. It can also cause testicular atrophy and is a reproductive toxicant. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation, and deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52908 Supplier Medchemexpress LLC Supplier No. HYW011215S10MG
Dihexyl phthalate-3,4,5,6-d4 is the deuterium labeled Dihexyl phthalate-3,4,5,6. Dihexyl phthalate is one of the commonly used phthalate esters in various plastics and consumer products. It is classified as a priority pollutant and an endocrine disruptor, capable of inducing reactive oxygen species (ROS) accumulation, promoting inflammation, and leading to increased apoptosis and inflammation-related gene expression levels. It can also cause testicular atrophy and is a reproductive toxicant. Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation, and deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.