Learn More
Medchemexpress LLC Dihexyl phthalate-3,4,5,6-d4 | 1015854-55-3 | 99.0% | 338.47 | 5 MG
SDP
Catalog No. 5000451885
Encompass_Preferred
Dihexyl phthalate-3,4,5,6-d4 is a deuterium labeled form of Dihexyl phthalate, a commonly used phthalate ester found in various plastics and consumer products. Classified as a priority pollutant and endocrine disruptor, it can induce reactive oxygen species (ROS) accumulation and promote inflammation. It is also known to cause testicular atrophy and is considered a reproductive toxicant.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
Catalog No. 50-004-51885 Supplier Medchemexpress LLC Supplier No. HYW011215S5MG
Dihexyl phthalate-3,4,5,6-d4 is a deuterium labeled form of Dihexyl phthalate, a commonly used phthalate ester found in various plastics and consumer products. Classified as a priority pollutant and endocrine disruptor, it can induce reactive oxygen species (ROS) accumulation and promote inflammation. It is also known to cause testicular atrophy and is considered a reproductive toxicant.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only.