Learn More
Medchemexpress LLC Dihexyl phthalate | 84-75-3 | 98.0% | 334.46 | 5 G
SDP
Catalog No. 5000451812
Encompass_Preferred
Dihexyl phthalate is a commonly used phthalate ester found in various plastics and consumer products. It is classified as a priority pollutant and an endocrine disruptor. It can induce reactive oxygen species (ROS) accumulation, promote inflammation, and significantly increase apoptosis and inflammation-related gene expression levels. Dihexyl phthalate can cause testicular atrophy and is considered a reproductive toxicant.
  • Induces reactive oxygen species (ROS) accumulation
  • Promotes inflammation
  • Increases apoptosis and inflammation-related gene expression levels
  • Can cause testicular atrophy
  • Considered a reproductive toxicant
Catalog No. 50-004-51812 Supplier Medchemexpress LLC Supplier No. HYW0112155G
Dihexyl phthalate is a commonly used phthalate ester found in various plastics and consumer products. It is classified as a priority pollutant and an endocrine disruptor. It can induce reactive oxygen species (ROS) accumulation, promote inflammation, and significantly increase apoptosis and inflammation-related gene expression levels. Dihexyl phthalate can cause testicular atrophy and is considered a reproductive toxicant.
  • Induces reactive oxygen species (ROS) accumulation
  • Promotes inflammation
  • Increases apoptosis and inflammation-related gene expression levels
  • Can cause testicular atrophy
  • Considered a reproductive toxicant