Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dihexylamine

Catalog No. 131202100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-120-2100ML 100 mL
13-120-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-120-2100ML Supplier Sigma Aldrich Supplier No. 131202100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dihexylamine nitrosation rate was enhanced when the reaction was carried out in the presence of micelles containing bile acid conjugates and lecithin.Dihexylamine was used in the preparation of succinic acid.

Specifications

CAS 143-16-8
Density 0.795 g/mL (at 25°C (literature))
Boiling Point 192°C to 195°C (lit.)
Molecular Formula C12H27N
Refractive Index n20/D 1.432 (literature)
Linear Formula [CH3(CH2)5]2 NH
MDL Number MFCD00009521
Quantity 100 mL
Molecular Weight (g/mol) 185.35
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.