Learn More
Medchemexpress LLC Dihomo-γ-linolenic acid-d6 | 81540-86-5 | 99.0% | C20H28D6O2 | 1 MG
SDP
Catalog No. 5000432448
Encompass_Preferred
Dihomo-γ-linolenic acid-d6 (DGLA-d6) is the deuterium labeled Dihomo-γ-linolenic acid. Dihomo-γ-linolenic acid (all-cis-8,11,14-Eicosatrienoic acid) is a 20-carbon ω-6 fatty acid, with anti-inflammatory and anti-proliferative activities. It attenuates atherosclerosis in the apolipoprotein E deficient mouse model system. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • 20-carbon ω-6 fatty acid
  • Anti-inflammatory activities
  • Anti-proliferative activities
  • Attenuates atherosclerosis
  • Can be used as a tracer
  • Used as an internal standard for quantitative analysis
Catalog No. 50-004-32448 Supplier Medchemexpress LLC Supplier No. HYA0143S1MG
Dihomo-γ-linolenic acid-d6 (DGLA-d6) is the deuterium labeled Dihomo-γ-linolenic acid. Dihomo-γ-linolenic acid (all-cis-8,11,14-Eicosatrienoic acid) is a 20-carbon ω-6 fatty acid, with anti-inflammatory and anti-proliferative activities. It attenuates atherosclerosis in the apolipoprotein E deficient mouse model system. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • 20-carbon ω-6 fatty acid
  • Anti-inflammatory activities
  • Anti-proliferative activities
  • Attenuates atherosclerosis
  • Can be used as a tracer
  • Used as an internal standard for quantitative analysis