Learn More
Cayman Chemical Dihomo-γ-Linolenic Acid ethyl ester, Purity: ≥98%, CAS: 55968-21-3, 100 mg
SDP
Catalog No. 502895467
Encompass_Preferred
An ester form of DGLA; dietary administration of ethyl DGLA at 1 g/kg per day increases PGE1 and PGE2 secretion in rabbit renal papilla; increases triglyceride levels and the proportion of 20:3ω6 and 20:4ω6 phospholipids of the renal medulla in rabbits
Catalog No. 50-289-5467 Supplier Cayman Chemical Supplier No. 90236100 MG
An ester form of DGLA; dietary administration of ethyl DGLA at 1 g/kg per day increases PGE1 and PGE2 secretion in rabbit renal papilla; increases triglyceride levels and the proportion of 20:3ω6 and 20:4ω6 phospholipids of the renal medulla in rabbits