Learn More
Cayman Chemical Dihomo-γ-Linolenic Acid ethyl ester, Purity: ≥98%, CAS: 55968-21-3, 50 mg
SDP
Catalog No. 502892995
Encompass_Preferred
An ester form of DGLA; dietary administration of ethyl DGLA at 1 g/kg per day increases PGE1 and PGE2 secretion in rabbit renal papilla; increases triglyceride levels and the proportion of 20:3ω6 and 20:4ω6 phospholipids of the renal medulla in rabbits
Catalog No. 50-289-2995 Supplier Cayman Chemical Supplier No. 9023650 MG
An ester form of DGLA; dietary administration of ethyl DGLA at 1 g/kg per day increases PGE1 and PGE2 secretion in rabbit renal papilla; increases triglyceride levels and the proportion of 20:3ω6 and 20:4ω6 phospholipids of the renal medulla in rabbits