Learn More
Cayman Chemical Dihomo-γ-Linolenic Acid, Purity: ≥98%, CAS: 1783-84-2, 10 mg
SDP
Catalog No. 502895049
Encompass_Preferred
An elongation product of γ-linolenic acid (18:3), rapidly metabolized by fatty acid desaturases to produce arachidonic acid (20:4); metabolized through the cyclooxygenase pathway to produce 1-series prostaglandins; can reduce atopic dermatitis and atherosclerosis when added to the diet of mice
Catalog No. 50-289-5049 Supplier Cayman Chemical Supplier No. 9023010 MG
An elongation product of γ-linolenic acid (18:3), rapidly metabolized by fatty acid desaturases to produce arachidonic acid (20:4); metabolized through the cyclooxygenase pathway to produce 1-series prostaglandins; can reduce atopic dermatitis and atherosclerosis when added to the diet of mice